Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R2D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PPP2R2D |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17417020
![]() |
Novus Biologicals
NBP17417020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174170
![]() |
Novus Biologicals
NBP174170 |
100 μL |
Each for $487.50
|
|
|||||
Description
PPP2R2D Polyclonal specifically detects PPP2R2D in Mouse samples. It is validated for Western Blot.Specifications
PPP2R2D | |
Polyclonal | |
Rabbit | |
Q925E7 | |
55844 | |
Synthetic peptides corresponding to the C terminal of Ppp2r2d. Immunizing peptide sequence RRDVTLEASRENSKPRASLKPRKVCTGGKRKKDEISVDSLDFNKKILHTA. | |
Primary | |
50 kDa |
Western Blot | |
Unconjugated | |
RUO | |
KIAA1541, MDS026, PP2A subunit B isoform B55-delta, PP2A subunit B isoform delta, PP2A subunit B isoform PR55-delta, PP2A subunit B isoform R2-delta, protein phosphatase 2, regulatory subunit B, delta, protein phosphatase 2, regulatory subunit B, delta isoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B deltaisoform | |
PPP2R2D | |
IgG | |
This product is specific to Subunit or Isoform: B delta isoform. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title