Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ PPP2R2D Monoclonal Antibody (GT577)
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527837
Description
Positive Control: mouse brain Predicted Reactivity: Human (100%), Rat (99%), Zebrafish (95%), Xenopus laevis (95%), Chicken (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PPP2R2D is the B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment. 
            Specifications
| PPP2R2D | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative | |
| Q66LE6, Q925E7 | |
| PPP2R2D | |
| Recombinant protein encompassing a sequence within the center region of Human PPP2R2D. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 | 
| Western Blot | |
| GT577 | |
| Unconjugated | |
| PPP2R2D | |
| 1300017E19Rik; D7Ertd753e; Kiaa1541; MDS026; PP2A subunit B isoform B55-delta; PP2A subunit B isoform delta; PP2A subunit B isoform PR55-delta; PP2A subunit B isoform R2-delta; PP2A, subunit B, B55-delta isoform; PP2A, subunit B, B-delta isoform; PP2A, subunit B, PR55-delta isoform; PP2A, subunit B, R2-delta isoform; PPP2R2D; protein phosphatase 2 regulatory subunit B, delta; protein phosphatase 2 regulatory subunit Bdelta; protein phosphatase 2, regulatory subunit B, delta; protein phosphatase 2, regulatory subunit B, delta isoform; protein phosphatase 2A B regulatory subunit delta isoform; protein phosphatase 2A, regulatory subunit B, delta isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform | |
| Mouse | |
| Protein G | |
| RUO | |
| 52432, 55844 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            