Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530763 Supplier Invitrogen™ Supplier No. PA530763
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32, mouse brain, rat brain. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (95%), Xenopus laevis (95%), Chicken (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PPP2R2D is the B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R2D
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PPP2R2D
Gene Accession No. P56932, Q66LE6, Q925E7
Gene Alias 1300017E19Rik; D7Ertd753e; Kiaa1541; MDS026; PP2A subunit B isoform B55-delta; PP2A subunit B isoform delta; PP2A subunit B isoform PR55-delta; PP2A subunit B isoform R2-delta; PP2A, subunit B, B55-delta isoform; PP2A, subunit B, B-delta isoform; PP2A, subunit B, PR55-delta isoform; PP2A, subunit B, R2-delta isoform; PPP2R2D; protein phosphatase 2 regulatory subunit B, delta; protein phosphatase 2 regulatory subunit Bdelta; protein phosphatase 2, regulatory subunit B, delta; protein phosphatase 2, regulatory subunit B, delta isoform; protein phosphatase 2A B regulatory subunit delta isoform; protein phosphatase 2A, regulatory subunit B, delta isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform
Gene Symbols PPP2R2D
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP2R2D. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246255, 52432, 55844
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.