Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS5759R

Description
Protein phosphatase 2 (formerly named type 2A) is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B′/PR61, and B′/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holoenzyme. The product of this gene belongs to the B′ family. The B′ family has been further divided into subfamilies. The product of this gene belongs to the alpha subfamily of regulatory subunit B′. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
PPP2R3 | |
Polyclonal | |
Unconjugated | |
PPP2R3A | |
3222402P14Rik; A730042E07; BOS_1271; DNA for thyroid hormone receptor binding site (258bp); LOC787607; PP2A subunit B isoform PR72/PR130; PP2A subunit B isoform R3 isoform; PP2A subunit B isoforms B72/B130; PP2A subunit B isoforms B′′-PR72/PR130; PP2A, subunit B, R3 isoform; PPP2R3; PPP2R3A; PR130; PR72; protein phosphatase 2 (formerly 2A), regulatory subunit B′′ (PR 72), alpha isoform and (PR 130), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B′′, alpha; protein phosphatase 2 regulatory subunit B′′, alpha; protein phosphatase 2 regulatory subunit B′′alpha; protein phosphatase 2, regulatory subunit B′′, alpha; Serine/threonine-protein phosphatase 2A 72/130 kDa regulatory subunit B; Serine/threonine-protein phosphatase 2A regulatory subunit B′′ subunit alpha; serine/threonine-protein phosphatase 2A regulatory subunit B′′ subunit alpha-like | |
Rabbit | |
Protein A | |
RUO | |
5523 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q06190 | |
PPP2R3A | |
KLH conjugated synthetic peptide derived from human PPP2R3 . | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction