Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530127 Supplier Invitrogen™ Supplier No. PA530127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, U87-MG, MCF-7. Predicted reactivity: Human (99%), Mouse (90%), Rat (90%), Rhesus Monkey (97%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Protein phosphatase 2 (formerly named type 2A) is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B'/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holoenzyme. The product of this gene belongs to the B' family. The B' family has been further divided into subfamilies. The product of this gene belongs to the alpha subfamily of regulatory subunit B'. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R3A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PPP2R3A
Gene Accession No. Q06190
Gene Alias 3222402P14Rik; A730042E07; DNA for thyroid hormone receptor binding site (258bp); PP2A subunit B isoform PR72/PR130; PP2A subunit B isoform R3 isoform; PP2A subunit B isoforms B72/B130; PP2A subunit B isoforms B''-PR72/PR130; PP2A, subunit B, R3 isoform; PPP2R3; PPP2R3A; PR130; PR72; protein phosphatase 2 (formerly 2A), regulatory subunit B'' (PR 72), alpha isoform and (PR 130), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B'', alpha; protein phosphatase 2 regulatory subunit B'', alpha; protein phosphatase 2 regulatory subunit B''alpha; protein phosphatase 2, regulatory subunit B'', alpha; serine/threonine-protein phosphatase 2A 72/130 kDa regulatory subunit B; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha-like protein
Gene Symbols PPP2R3A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PPP2R3A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235542, 363122, 5523
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.