Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550734 Supplier Invitrogen™ Supplier No. PA550734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PPP2R3A protein.

Protein phosphatase 2 (formerly named type 2A) is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B'/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holoenzyme. The product of this gene belongs to the B' family. The B' family has been further divided into subfamilies. The product of this gene belongs to the alpha subfamily of regulatory subunit B'. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PPP2R3A
Gene Accession No. Q06190
Gene Alias 3222402P14Rik; A730042E07; DNA for thyroid hormone receptor binding site (258bp); PP2A subunit B isoform PR72/PR130; PP2A subunit B isoform R3 isoform; PP2A subunit B isoforms B72/B130; PP2A subunit B isoforms B''-PR72/PR130; PP2A, subunit B, R3 isoform; PPP2R3; PPP2R3A; PR130; PR72; protein phosphatase 2 (formerly 2A), regulatory subunit B'' (PR 72), alpha isoform and (PR 130), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B'', alpha; protein phosphatase 2 regulatory subunit B'', alpha; protein phosphatase 2 regulatory subunit B''alpha; protein phosphatase 2, regulatory subunit B'', alpha; serine/threonine-protein phosphatase 2A 72/130 kDa regulatory subunit B; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha-like protein
Gene Symbols PPP2R3A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human protein phosphatase 2, regulatory subunit B'', alpha.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5523
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.