Learn More
Invitrogen™ PPP2R3B Monoclonal Antibody (2E12)

Description
Peptide Sequence: MIDRIFSGAV TRGRKVQKEG KISYADFVWF LISEEDKKTP TSIEYWFRCM DLDGDGALSM FELEYFYEEQ CRRLDSMAIE ALPFQDCLCQ MLDLVKPRTE GKITLQDLKR CKLANVFFDT FFNIEKYLDH EQKEQISLLR DGDSGGPELS DWEKYAAEEY DILVAEETAG EPWEDGFEAE LSPVEQKLSA LRSPLAQRPF FEAPSPLGAV DLYEYACGDE DLEPL.
Specifications
Specifications
| Antigen | PPP2R3B |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2E12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PPP2R3B |
| Gene Accession No. | Q9Y5P8 |
| Gene Alias | NYREN8; NY-REN-8; NY-REN-8 antigen; PP2A subunit B isoform PR48; PP2A, subunit B, PR48 isoform; PPP2R3B; PPP2R3L; PPP2R3LY; PR48; protein phosphatase 2 (formerly 2A), regulatory subunit B' ', beta; protein phosphatase 2 regulatory subunit B' ', beta; protein phosphatase 2 regulatory subunit B' 'beta; protein phosphatase 2, regulatory subunit B' ', beta; Protein phosphatase 2A 48 kDa regulatory subunit; serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B; serine/threonine-protein phosphatase 2A regulatory subunit B' ' subunit beta |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.