Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599073 Supplier Invitrogen™ Supplier No. PA599073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP2R3B Polyclonal Antibody for ELISA, IHC (P), Western Blot

Protein phosphatase 2 is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B' '/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B' ' family. The B' ' family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B' '. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R3B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP2R3B
Gene Accession No. Q9Y5P8
Gene Alias NYREN8; NY-REN-8; NY-REN-8 antigen; PP2A subunit B isoform PR48; PP2A, subunit B, PR48 isoform; PPP2R3B; PPP2R3L; PPP2R3LY; PR48; protein phosphatase 2 (formerly 2A), regulatory subunit B' ', beta; protein phosphatase 2 regulatory subunit B' ', beta; protein phosphatase 2 regulatory subunit B' 'beta; protein phosphatase 2, regulatory subunit B' ', beta; Protein phosphatase 2A 48 kDa regulatory subunit; serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B; serine/threonine-protein phosphatase 2A regulatory subunit B' ' subunit beta
Gene Symbols PPP2R3B
Host Species Rabbit
Immunogen Recombinant Human Serine/threonine-protein phosphatase 2A regulatory subunit B' ' subunit beta protein (356-575AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28227
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.