Learn More
PPP2R3B Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300965A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Protein phosphatase 2 is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B′/PR61, and B′′/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B′′ family. The B′′ family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B′′. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| PPP2R3B | |
| Polyclonal | |
| Unconjugated | |
| PPP2R3B | |
| Serine/threonine-protein phosphatase 2A regulatory subunit B′′ subunit beta; Protein phosphatase 2A 48 kDa regulatory subunit; PP2A, subunit B, PR48 isoform; PPP2R3B; PPP2R3L; NY-REN-8; PPP2R3LY | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28227 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9Y5P8 | |
| PPP2R3B | |
| Between 300 and 350 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.