Learn More
Invitrogen™ PPP2R3C Polyclonal Antibody

Description
The antibody detects endogenous levels of total PPP2R3C protein.
Specifications
Specifications
| Antigen | PPP2R3C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Ppp2r3c |
| Gene Accession No. | Q969Q6 |
| Gene Alias | 4930511A21Rik; 5730412A08Rik; C14orf10; chromosome 14 open reading frame 10; G4-1; G5pr; hypothetical protein LOC533147; MNCb-1932; phosphatase subunit gene g4-1; Ppp2r3c; protein phosphatase 2 (formerly 2A), regulatory subunit B''; protein phosphatase 2 (formerly 2A), regulatory subunit B'', gamma; protein phosphatase 2 regulatory subunit B'', gamma; protein phosphatase 2 regulatory subunit B''gamma; protein phosphatase 2, regulatory subunit B'', gamma; protein phosphatase subunit G5PR; RGD1309207; rhabdomyosarcoma antigen MU-RMS-40.6A; Rhabdomyosarcoma antigen MU-RMS-40.6A/6C; rhabdomyosarcoma antigen Mu-RMS-40.6C; serine/threonine-protein phosphatase 2A regulatory subunit B'' gamma subunit; serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma; similar to phosphatase subunit gene g4-1; zgc:55473 |
| Gene Symbols | Ppp2r3c |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.