Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP2R3C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566214
Description
Immunogen sequence: LATPNTCPNKK KSEQELKDEE MDLFTKYYSE WKGGRKNTNE FYKTIPRFYY RLPAEDEVLL QKLREESRAV FLQRKSRELL DNEELQNLWF LLDKHQTPPM IGEEAMINYE NF Highest antigen sequence identity to the following orthologs - mouse 96%, rat 95%.
P2R3C may regulate MCM3AP phosphorylation through phosphatase recruitment. It may play a role in the activation-induced cell death of B-cells.
Specifications
PPP2R3C | |
Polyclonal | |
Unconjugated | |
Ppp2r3c | |
4930511A21Rik; 5730412A08Rik; C14orf10; chromosome 14 open reading frame 10; G4-1; G5pr; hypothetical protein LOC533147; MNCb-1932; phosphatase subunit gene g4-1; Ppp2r3c; protein phosphatase 2 (formerly 2A), regulatory subunit B''; protein phosphatase 2 (formerly 2A), regulatory subunit B'', gamma; protein phosphatase 2 regulatory subunit B'', gamma; protein phosphatase 2 regulatory subunit B''gamma; protein phosphatase 2, regulatory subunit B'', gamma; protein phosphatase subunit G5PR; RGD1309207; rhabdomyosarcoma antigen MU-RMS-40.6A; Rhabdomyosarcoma antigen MU-RMS-40.6A/6C; rhabdomyosarcoma antigen Mu-RMS-40.6C; serine/threonine-protein phosphatase 2A regulatory subunit B'' gamma subunit; serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma; similar to phosphatase subunit gene g4-1; zgc:55473 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55012 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969Q6 | |
Ppp2r3c | |
Recombinant Human PPP2R3C. Recombinant protein control fragment (Product #RP-106895). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction