Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598656 Supplier Invitrogen™ Supplier No. PA598656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP2R3C Polyclonal Antibody for Western Blot, IHC (P), ELISA

P2R3C may regulate MCM3AP phosphorylation through phosphatase recruitment. It may play a role in the activation-induced cell death of B-cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R3C
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ppp2r3c
Gene Accession No. Q969Q6, Q9JK24
Gene Alias 4930511A21Rik; 5730412A08Rik; C14orf10; chromosome 14 open reading frame 10; G4-1; G5pr; hypothetical protein LOC533147; MNCb-1932; phosphatase subunit gene g4-1; Ppp2r3c; protein phosphatase 2 (formerly 2A), regulatory subunit B; protein phosphatase 2 (formerly 2A), regulatory subunit B, gamma; protein phosphatase 2 regulatory subunit B, gamma; protein phosphatase 2 regulatory subunit Bgamma; protein phosphatase 2, regulatory subunit B, gamma; protein phosphatase subunit G5PR; RGD1309207; rhabdomyosarcoma antigen MU-RMS-40.6A; Rhabdomyosarcoma antigen MU-RMS-40.6A/6C; rhabdomyosarcoma antigen Mu-RMS-40.6C; serine/threonine-protein phosphatase 2A regulatory subunit B gamma subunit; serine/threonine-protein phosphatase 2A regulatory subunit B subunit gamma; similar to phosphatase subunit gene g4-1; zgc:55473
Gene Symbols Ppp2r3c
Host Species Rabbit
Immunogen Recombinant Human Serine/threonine-protein phosphatase 2A regulatory subunit B subunit gamma protein (1-160AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55012, 59032
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.