Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R4 Antibody (4D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237530
Description
PPP2R4 Monoclonal specifically detects PPP2R4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
PPP2R4 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
EC 5.2.1.8, MGC2184, Phosphotyrosyl phosphatase activator, PP2A, PP2A phosphatase activator, PP2A subunit B' isoform PR53, PP2A, subunit B', PR53 isoform, protein phosphatase 2A activator, regulatory subunit 4, regulatory subunit B' (PR 53), serine/threonine-protein phosphatase 2A activator, Serine/threonine-protein phosphatase 2A regulatory subunit 4, Serine/threonine-protein phosphatase 2A regulatory subunit B' | |
Mouse | |
41 kDa | |
0.1 mL | |
Protein Phosphatase | |
5524 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
4D9 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
PPP2R4 | |
Purified recombinant fragment of human PPP2R4 expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction