Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP2R4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528296
Description
Recommended positive controls: 293T, A431, mouse brain. Predicted reactivity: Mouse (96%), Rat (94%), Rabbit (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Protein phosphatase 2A is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2A holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B''/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B' family. This gene encodes a specific phosphotyrosyl phosphatase activator of the dimeric form of protein phosphatase 2A. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
PPP2R4 | |
Polyclonal | |
Unconjugated | |
PTPA | |
2610042B21Rik; C77440; MGC2184; N28142; OTTHUMP00000022333; phosphotyrosyl phosphatase activator; PP2A; PP2A phosphatase activator; PP2A subunit B' isoform PR53; PP2A, subunit B', PR53 isoform; Ppp2r4; PR53; protein phosphatase 2 phosphatase activator; protein phosphatase 2 regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit B; protein phosphatase 2A regulatory subunit 4; protein phosphatase 2A, regulatory subunit B (PR 53); protein phosphatase 2A, regulatory subunit B' (PR 53); PTPA; Serine/threonine-protein phosphatase 2A activator; serine/threonine-protein phosphatase 2A regulatory subunit 4; Serine/threonine-protein phosphatase 2A regulatory subunit B' | |
Rabbit | |
Protein A | |
RUO | |
110854, 5524 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
6.31 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P58389, Q15257 | |
PTPA | |
Recombinant protein encompassing a sequence within the center region of human PPP2R4. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction