Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529755 Supplier Invitrogen™ Supplier No. PA529755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (96%), Rat (95%), Rabbit (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Protein phosphatase 2A is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2A holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B''/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B' family. This gene encodes a specific phosphotyrosyl phosphatase activator of the dimeric form of protein phosphatase 2A. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PTPA
Gene Accession No. Q15257
Gene Alias 2610042B21Rik; C77440; MGC2184; N28142; OTTHUMP00000022333; phosphotyrosyl phosphatase activator; PP2A; PP2A phosphatase activator; PP2A subunit B' isoform PR53; PP2A, subunit B', PR53 isoform; Ppp2r4; PR53; protein phosphatase 2 phosphatase activator; protein phosphatase 2 regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit B; protein phosphatase 2A regulatory subunit 4; protein phosphatase 2A, regulatory subunit B (PR 53); protein phosphatase 2A, regulatory subunit B' (PR 53); PTPA; Serine/threonine-protein phosphatase 2A activator; serine/threonine-protein phosphatase 2A regulatory subunit 4; Serine/threonine-protein phosphatase 2A regulatory subunit B'
Gene Symbols PTPA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP2R4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5524
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.