Learn More
Description
Specifications
Specifications
| Antigen | PPP2R5A |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
| Formulation | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Gene Alias | B56A, MGC131915, PP2A B subunit isoform B56-alpha, PP2A B subunit isoform B'-alpha, PP2A B subunit isoform PR61-alpha, PP2A B subunit isoform R5-alpha, PP2A, B subunit, B' alpha isoform, PP2A, B subunit, B56 alpha isoform, PP2A, B subunit, PR61 alpha isoform, PP2A, B subunit, R5 alpha isoform, PR61A, PR61alpha, protein phosphatase 2, regulatory subunit B (B56), alpha isoform, protein phosphatase 2, regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alphaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform |
| Gene Symbols | PPP2R5A |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TLNELVEYVSTNRGVIVESAYSD |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
