Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R5B Polyclonal Antibody

Goat Polyclonal Antibody

Supplier:  Invitrogen™ PA131465

Catalog No. PIPA131465


Only null left
Add to Cart

Description

Description

Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX21366-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPP2R5B
Polyclonal
Unconjugated
PPP2R5B
B56B; B'beta; BC026670; FLJ35411; LOC100909468; PP2A; PP2A B subunit isoform B56-beta; PP2A B subunit isoform B'-beta; PP2A B subunit isoform PR61-beta; PP2A B subunit isoform R5-beta; Ppp2r5b; PR61B; protein phosphatase 2 regulatory subunit B', beta; protein phosphatase 2 regulatory subunit B'beta; protein phosphatase 2, regulatory subunit B (B56), beta isoform; protein phosphatase 2, regulatory subunit B', beta; protein phosphatase 2, regulatory subunit B', beta isoform; protein phosphotase 2, regulatory subunit B (B56), beta isoform; rCG_47223; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, beta isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Goat
Antigen Affinity Chromatography
RUO
5526
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q15173
PPP2R5B
Synthetic peptide corresponding to amino acids CQRLTPQVAASGGQS from C-Terminus of Human PPP2R5B (aa 484-497).
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.