Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557740 Supplier Invitrogen™ Supplier No. PA557740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKLFDELTAS YKLEKQQEQQ KAQERQELWQ GLEELRLRRL QGTQGAKEAP LQRLTP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP2R5B
Gene Accession No. Q15173, Q6PD28, Q80W83
Gene Alias B56B; B'beta; BC026670; FLJ35411; LOC100909468; PP2A; PP2A B subunit isoform B56-beta; PP2A B subunit isoform B'-beta; PP2A B subunit isoform PR61-beta; PP2A B subunit isoform R5-beta; Ppp2r5b; PR61B; protein phosphatase 2 regulatory subunit B', beta; protein phosphatase 2 regulatory subunit B'beta; protein phosphatase 2, regulatory subunit B (B56), beta isoform; protein phosphatase 2, regulatory subunit B', beta; protein phosphatase 2, regulatory subunit B', beta isoform; protein phosphotase 2, regulatory subunit B (B56), beta isoform; rCG_47223; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, beta isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Gene Symbols PPP2R5B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP2R5B. Recombinant protein control fragment (Product #RP-97258).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225849, 309179, 5526
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.