Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP2R5B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585443
Description
Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (100%), Rabbit (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.
Specifications
PPP2R5B | |
Polyclonal | |
Unconjugated | |
PPP2R5B | |
B56B; B'beta; BC026670; FLJ35411; LOC100909468; PP2A; PP2A B subunit isoform B56-beta; PP2A B subunit isoform B'-beta; PP2A B subunit isoform PR61-beta; PP2A B subunit isoform R5-beta; Ppp2r5b; PR61B; protein phosphatase 2 regulatory subunit B', beta; protein phosphatase 2 regulatory subunit B'beta; protein phosphatase 2, regulatory subunit B (B56), beta isoform; protein phosphatase 2, regulatory subunit B', beta; protein phosphatase 2, regulatory subunit B', beta isoform; protein phosphotase 2, regulatory subunit B (B56), beta isoform; rCG_47223; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, beta isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.92 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q15173 | |
PPP2R5B | |
Recombinant protein encompassing a sequence within the center region of human PPP2R5B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction