Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582984 Supplier Invitrogen™ Supplier No. PA582984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRNSKTHWNK TIHGLIYNAL KLFMEMNQKL FDDCTQQFKA EKLKEKLKMK EREEAWVKIE NLAKANPQAQ KDPKKDRPLA RRKSELPQDP HTKKALEAHC Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 70%.

Phosphatases are involved in the regulation of MAP kinases.Protein phosphatase 2A (PP2A) is a critical component of the pathway responsible for p38 MAPK up-regulation of SERT catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP2R5C
Gene Accession No. Q13362, Q60996
Gene Alias 2610043M05Rik; 2700063L20Rik; AI060890; AW545884; B' alpha regulatory subunit; B55-GAMMA; B56/PP2A gamma; B56G; Band 8A; C85228; D12Bwg0916e; gamma isoform of regulatory subunit B56, protein phosphatase 2A; IMYPNO; IMYPNO1; KIAA0044; MGC23064; MGC33570; mKIAA0044; PP2A B subunit isoform B56-gamma; PP2A B subunit isoform B'-alpha-3; PP2A B subunit isoform B'-gamma; PP2A B subunit isoform PR61-gamma; PP2A B subunit isoform R5-gamma; Ppp2r5c; PR52; PR55G; PR61G; protein phosphatase 2 regulatory subunit B', gamma; protein phosphatase 2 regulatory subunit B'gamma; protein phosphatase 2, regulatory subunit B (B56), gamma isoform; protein phosphatase 2, regulatory subunit B', gamma; protein phosphatase 2, regulatory subunit B', gamma isoform; renal carcinoma antigen NY-REN-29; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform
Gene Symbols PPP2R5C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP2R5C. Recombinant protein control fragment (Product #RP-88937).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26931, 5527, 691318
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.