Learn More
Invitrogen™ PPP2R5C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110209
Description
Immunogen sequence: MLTCNKAGSR MVVDAANSNG PFQPVVLLHI RDVPPADQEK LFIQKLRQCC VLFDFVSDPL SDLKWKEVKR AALSEMVEYI THNRNVITEP IYPEVVHMFA VNMFRTLPPS SNPTGAEFDP EEDEPTLEAA WPHLQLVYEF FLRFLESPDF QPNIAKKYID QKFVLQLLEL FDSEDPRERD FLKTTLHRIY GKFLGLRAYI RKQINNIFYR FIYETEHHNG IAELLEILGS IINGFALPLK EEHKIFLLKV LLPLHKVKSL SVYHPQLAYC VVQFLEKDST LTEPVVMALL KYWPKTHSPK EVMFLNELEE ILDVIEPSEF.
Phosphatases are involved in the regulation of MAP kinases.Protein phosphatase 2A (PP2A) is a critical component of the pathway responsible for p38 MAPK up-regulation of SERT catalytic activity.
Specifications
| PPP2R5C | |
| Polyclonal | |
| Unconjugated | |
| PPP2R5C | |
| 2610043M05Rik; 2700063L20Rik; AI060890; AW545884; B' alpha regulatory subunit; B55-GAMMA; B56/PP2A gamma; B56G; Band 8A; C85228; D12Bwg0916e; gamma isoform of regulatory subunit B56, protein phosphatase 2A; IMYPNO; IMYPNO1; KIAA0044; MGC23064; MGC33570; mKIAA0044; PP2A B subunit isoform B56-gamma; PP2A B subunit isoform B'-alpha-3; PP2A B subunit isoform B'-gamma; PP2A B subunit isoform PR61-gamma; PP2A B subunit isoform R5-gamma; Ppp2r5c; PR52; PR55G; PR61G; protein phosphatase 2 regulatory subunit B', gamma; protein phosphatase 2 regulatory subunit B'gamma; protein phosphatase 2, regulatory subunit B (B56), gamma isoform; protein phosphatase 2, regulatory subunit B', gamma; protein phosphatase 2, regulatory subunit B', gamma isoform; renal carcinoma antigen NY-REN-29; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26931, 5527, 691318 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13362, Q60996 | |
| PPP2R5C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human PP2A-B56?/PR61?/PP2A-B56?/PR61?/PPP2R5C (NP_8487021). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.