Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115789
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115789 Supplier Invitrogen™ Supplier No. PA5115789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP2R5C.

Phosphatases are involved in the regulation of MAP kinases.Protein phosphatase 2A (PP2A) is a critical component of the pathway responsible for p38 MAPK up-regulation of SERT catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PPP2R5C
Gene Accession No. Q13362
Gene Alias 2610043M05Rik; 2700063L20Rik; AI060890; AW545884; B' alpha regulatory subunit; B55-GAMMA; B56/PP2A gamma; B56G; Band 8A; C85228; D12Bwg0916e; gamma isoform of regulatory subunit B56, protein phosphatase 2A; IMYPNO; IMYPNO1; KIAA0044; MGC23064; MGC33570; mKIAA0044; PP2A B subunit isoform B56-gamma; PP2A B subunit isoform B'-alpha-3; PP2A B subunit isoform B'-gamma; PP2A B subunit isoform PR61-gamma; PP2A B subunit isoform R5-gamma; Ppp2r5c; PR52; PR55G; PR61G; protein phosphatase 2 regulatory subunit B', gamma; protein phosphatase 2 regulatory subunit B'gamma; protein phosphatase 2, regulatory subunit B (B56), gamma isoform; protein phosphatase 2, regulatory subunit B', gamma; protein phosphatase 2, regulatory subunit B', gamma isoform; renal carcinoma antigen NY-REN-29; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform
Gene Symbols PPP2R5C
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP2R5C(Accession Q13362), corresponding to amino acid residues M452-D502.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5527, 691318
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.