Learn More
Invitrogen™ PPP2R5C Polyclonal Antibody

Description
PPP2R5C Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PPP2R5C |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | PPP2R5C |
| Gene Accession No. | Q13362, Q60996 |
| Gene Alias | 2610043M05Rik; 2700063L20Rik; AI060890; AW545884; B' alpha regulatory subunit; B55-GAMMA; B56/PP2A gamma; B56G; Band 8A; C85228; D12Bwg0916e; gamma isoform of regulatory subunit B56, protein phosphatase 2A; IMYPNO; IMYPNO1; KIAA0044; MGC23064; MGC33570; mKIAA0044; PP2A B subunit isoform B56-gamma; PP2A B subunit isoform B'-alpha-3; PP2A B subunit isoform B'-gamma; PP2A B subunit isoform PR61-gamma; PP2A B subunit isoform R5-gamma; Ppp2r5c; PR52; PR55G; PR61G; protein phosphatase 2 regulatory subunit B', gamma; protein phosphatase 2 regulatory subunit B'gamma; protein phosphatase 2, regulatory subunit B (B56), gamma isoform; protein phosphatase 2, regulatory subunit B', gamma; protein phosphatase 2, regulatory subunit B', gamma isoform; renal carcinoma antigen NY-REN-29; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform |
| Gene Symbols | PPP2R5C |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.