Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R5D Antibody (OTI5E7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273587B
Description
PPP2R5D Monoclonal specifically detects PPP2R5D in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PPP2R5D | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PPP2R5D | |
Full length recombinant protein of human PPP2R5D (NP_006236) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI5E7 | |
Western Blot, Immunohistochemistry | |
MGC2134, MGC8949, PP2A B subunit isoform B56-delta, PP2A B subunit isoform B′-delta, PP2A B subunit isoform PR61-delta, PP2A B subunit isoform R5-delta, PP2A, B subunit, B′ delta isoform, PP2A, B subunit, B56 delta isoform, PP2A, B subunit, PR61 delta isoform, PP2A, B subunit, R5 delta isoform, protein phosphatase 2, regulatory subunit B′, delta, regulatory subunit B (B56), delta isoform, regulatory subunit B′, delta isoform, Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, deltaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform | |
Mouse | |
Affinity Purified | |
Neuroscience, Wnt Signaling Pathway | |
5528.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction