Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R5E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PPP2R5E |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17417320
![]() |
Novus Biologicals
NBP17417320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174173
![]() |
Novus Biologicals
NBP174173 |
100 μL |
Each for $487.50
|
|
|||||
Description
PPP2R5E Polyclonal specifically detects PPP2R5E in Mouse samples. It is validated for Western Blot.Specifications
PPP2R5E | |
Polyclonal | |
Rabbit | |
Q61151 | |
5529 | |
Synthetic peptides corresponding to the N terminal of Ppp2r5e. Immunizing peptide sequence SCNIFRTLPPSDSNEFDPEEDEPTLEASWPHLQLVYEFFIRFLESQEFQP. | |
Primary | |
51 kDa |
Western Blot | |
Unconjugated | |
RUO | |
epsilon isoform of regulatory subunit B56, protein phosphatase 2A, PP2A B subunit isoform B56-epsilon, PP2A B subunit isoform B'-epsilon, PP2A B subunit isoform PR61-epsilon, PP2A B subunit isoform R5-epsilon, PP2A, B subunit, B' epsilon, PP2A, B subunit, B56 epsilon, PP2A, B subunit, PR61 epsilon, PP2A, B subunit, R5 epsilon, protein phosphatase 2, regulatory subunit B (B56), epsilon isoform, protein phosphatase 2, regulatory subunit B', epsilon isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, epsilon, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilonisoform | |
PPP2R5E | |
IgG | |
This product is specific to Subunit or Isoform: epsilon isoform. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title