Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R5E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517981
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517981 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517981 Supplier Invitrogen™ Supplier No. PA517981
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an epsilon isoform of the regulatory subunit B56 subfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5E
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PPP2R5E
Gene Accession No. Q16537, Q61151
Gene Alias 4633401M22Rik; AI449017; B56 epsilon isoform; B56beta; epsilon isoform of regulatory subunit B56, protein phosphatase 2A; Kiaa4006; PP2A B subunit isoform B56-epsilon; PP2A B subunit isoform B'-epsilon; PP2A B subunit isoform PR61-epsilon; PP2A B subunit isoform R5-epsilon; PP2A, B subunit, B' epsilon; PP2A, B subunit, B56 epsilon; PP2A, B subunit, PR61 epsilon; PP2A, B subunit, R5 epsilon; Ppp2r5e; protein phosphatase 2 regulatory subunit B', epsilon; protein phosphatase 2 regulatory subunit B'epsilon; protein phosphatase 2, regulatory subunit B (B56), epsilon isoform; protein phosphatase 2, regulatory subunit B', epsilon; protein phosphatase 2, regulatory subunit B', epsilon isoform; protein phosphatase 2A subunit beta; R5 epsilon isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, epsilon; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform
Gene Symbols PPP2R5E
Host Species Goat
Immunogen Synthetic peptide sequence (LKRGLRRDGIIPT) corresponding to the C-terminus amino acids of PPP2R5E (aa 455-467).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26932, 299147, 5529
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.