Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R5E Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989894
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9894 Supplier Bioss Supplier No. BS19971R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an epsilon isoform of the regulatory subunit B56 subfamily.

Specifications

Antigen PPP2R5E
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene PPP2R5E
Gene Accession No. Q16537
Gene Alias 4633401M22Rik; AI449017; B56 epsilon isoform; B56beta; epsilon isoform of regulatory subunit B56, protein phosphatase 2A; Kiaa4006; PP2A B subunit isoform B56-epsilon; PP2A B subunit isoform B′-epsilon; PP2A B subunit isoform PR61-epsilon; PP2A B subunit isoform R5-epsilon; PP2A, B subunit, B′ epsilon; PP2A, B subunit, B56 epsilon; PP2A, B subunit, PR61 epsilon; PP2A, B subunit, R5 epsilon; Ppp2r5e; protein phosphatase 2 regulatory subunit B′, epsilon; protein phosphatase 2 regulatory subunit B′epsilon; protein phosphatase 2, regulatory subunit B (B56), epsilon isoform; protein phosphatase 2, regulatory subunit B′, epsilon; protein phosphatase 2, regulatory subunit B′, epsilon isoform; protein phosphatase 2A subunit beta; R5 epsilon isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, epsilon; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform
Gene Symbols PPP2R5E
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PPP2R5E.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5529
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.