Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP3CA Mouse anti-Human, Rat, Clone: OTI5C6, liquid, TrueMAB™
SDP

Catalog No. 501683507
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3507 Supplier OriGene Supplier No. TA810584
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin. Dephosphorylates DNM1L, HSPB1 and SSH1.

Specifications

Antigen PPP3CA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PPP3CA
Gene Accession No. P63329, Q08209
Gene Alias CALN, CALNA, CALNA1, CCN1, CNA1, IECEE, IECEE1, PPP2B
Gene Symbols Ppp3ca
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 2-105 of human PPP3CA produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24674, 5530
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.