Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP3CB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100307 Supplier Invitrogen™ Supplier No. PA5100307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP3CB.

PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP3CB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP3CB
Gene Accession No. P16298, P48453
Gene Alias 1110063J16Rik; calcineurin A; calcineurin A beta; Calcineurin A protein; calcineurin A2; Calcineurin subunit A beta; calmodulin-dependent calcineurin A subunit beta isoform; CALNA2; Calnb; CAM-PRP catalytic subunit; CaN Abeta; CNA beta; CNA2; Cnab; CnAbeta; hypothetical protein LOC550554; PP2BA beta; PP2Bbeta; Ppp3cb; protein phosphatase 2B, catalytic subunit, beta isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform; protein phosphatase 3 catalytic subunit beta; protein phosphatase 3, catalytic subunit, beta isoform; protein phosphatase 3, catalytic subunit, beta isozyme; protein phosphatase from PCR fragment H32; protein phosphatase-2Bb; RP11-345K20.1; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit alpha; wu:fq19c08; zgc:110141
Gene Symbols PPP3CB
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP3CB, corresponding to a region within C-terminal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19056, 5532
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.