Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP3CB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570325
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.
Specifications
PPP3CB | |
Polyclonal | |
Unconjugated | |
PPP3CB | |
1110063J16Rik; calcineurin A; calcineurin A beta; Calcineurin A protein; calcineurin A2; Calcineurin subunit A beta; calmodulin-dependent calcineurin A subunit beta isoform; CALNA2; Calnb; CAM-PRP catalytic subunit; CaN Abeta; CNA beta; CNA2; Cnab; CnAbeta; hypothetical protein LOC550554; PP2BA beta; PP2Bbeta; Ppp3cb; protein phosphatase 2B, catalytic subunit, beta isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform; protein phosphatase 3 catalytic subunit beta; protein phosphatase 3, catalytic subunit, beta isoform; protein phosphatase 3, catalytic subunit, beta isozyme; protein phosphatase from PCR fragment H32; protein phosphatase-2Bb; RP11-345K20.1; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; serine/threonine-protein phosphatase 2B catalytic subunit alpha; wu:fq19c08; zgc:110141 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
24675 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P20651 | |
PPP3CB | |
synthetic peptide directed towards the middle region of rat PPP3CB. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction