Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP3CC Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145072 Supplier Invitrogen™ Supplier No. PA5145072
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RAHEAQDAGY RMYRKSQATG FPSLITIFSA PNYLDVYN Antigen sequence identity: Mouse 97%; Rat 97%.

Calmodulin-dependent protein phosphatase, calcineurin, is involved in a wide range of biologic activities, acting as a Ca(2+)-dependent modifier of phosphorylation status. In testis, the motility of the sperm is thought to be controlled by cAMP-dependent phosphorylation and a unique form of calcineurin appears to be associated with the flagellum. The calcineurin holoenzyme is composed of catalytic and regulatory subunits of 60 and 18 kD, respectively. At least 3 genes, calcineurin A-alpha, calcineurin A-beta, and calcineurin A-gamma (CALNA3), have been cloned for the catalytic subunit. These genes have been identified in humans, mice, and rats, and are highly conserved between species (90 to 95% amino acid identity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP3CC
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP3CC
Gene Accession No. P48454
Gene Alias Calcineurin A gamma; Calcineurin, testis-specific catalytic subunit; calmodulin-dependent calcineurin A subunit gamma isoform; CALNA3; Calnc; CAM-PRP catalytic subunit; CNA3; PP2BA gamma; PP2Bgamma; PPP3CC; protein phosphatase 2B, catalytic subunit, gamma isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, gamma isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, gamma isoform (calcineurin A gamma); protein phosphatase 3 catalytic subunit gamma; protein phosphatase 3, catalytic subunit, gamma isoform; protein phosphatase 3, catalytic subunit, gamma isozyme; serine/threonine-protein phosphatase 2B catalytic subunit gamma isoform
Gene Symbols PPP3CC
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 275-312. Recombinant protein control fragment (Product #a href=.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5533
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.