Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP3R1 Mouse anti-Bovine, Human, Rat, Clone: CN-A1, Abnova™

Catalog No. 89934797 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-797 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-797 Supplier Abnova Corporation Supplier No. MAB12449
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length native of bovine PPP3R1.

Specifications

Antigen PPP3R1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone CN-A1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native of bovine PPP3R1.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In ascites (1.2% sodium acetate, 2mg BSA, 0.01mg sodium azide)
Gene Symbols PPP3R1
Host Species Mouse
Immunogen Native purified PPP3R1 from bovine brain.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282321
Target Species Bovine, Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.