Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP3R1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | PPP3R1 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8574
![]()
|
Proteintech Group Inc
132101AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Protein phosphatase 3 (PPP3, formerly PP2B, Calcineurin), a serine/ threonine protein phosphatase, is a heterodimer composed of one catalytic subunit (PPP3C, Calcineurin A) and one regulatory subunit (PPP3R, Calcineurin B). PPP3R has two isoforms, PPP3R1 and PPP3R2. PPP3Rl is ubiquitously expressed in different tissues and is coded by the PPP3R1 gene which was mapped recently to human chromosome 2. PPP3Rl has four functional calcium-binding sites.Specifications
| PPP3R1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P63098 | |
| 5534 | |
| PPP3R1 Fusion Protein Ag3992 | |
| Primary | |
| -20°C | |
| PPP3R1 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Calcineurin subunit B type 1, CALNB1, CNA2, CNB, CNB1, PPP3R1 | |
| PPP3R1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title