Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP3R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103671 Supplier Invitrogen™ Supplier No. PA5103671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP3R2.

This gene encodes a member of the prolyl 3-hydroxylase subfamily of 2-oxo-glutarate-dependent dioxygenases. These enzymes play a critical role in collagen chain assembly, stability and cross-linking by catalyzing post-translational 3-hydroxylation of proline residues. Mutations in this gene are associated with nonsyndromic severe myopia with cataract and vitreoretinal degeneration, and downregulation of this gene may play a role in breast cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP3R2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP3R2
Gene Accession No. P28470, Q63811, Q96LZ3
Gene Alias calcineurin B subunit; calcineurin B, type II; calcineurin B, type II (19kDa); Calcineurin BII; calcineurin B-like protein; calcineurin subunit B type 2; CaNB2; CBLP; CnB2; CNBII; PP2B beta 2; PPP3R1-like; Ppp3r2; PPP3RL; protein phospatase 3, regulatory subunit B, alpha isoform (calcineurin B, type II); protein phospatase 3, regulatory subunit B, alpha isoform,type 2; protein phosphatase 2B regulatory subunit 2; protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), beta isoform (calcineurin B, type II); protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, beta isoform (calcineurin B, type II); protein phosphatase 3 (formerly 2B), regulatory subunit B, beta isoform; protein phosphatase 3 regulatory subunit B beta isoform; protein phosphatase 3 regulatory subunit B, beta; protein phosphatase 3, regulatory subunit B, alpha isoform (calcineurin B, type II); protein phosphatase 3, regulatory subunit B, beta; protein phosphatase 3, regulatory subunit B, beta isoform; ty
Gene Symbols PPP3R2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP3R2(Accession Q96LZ3), corresponding to amino acid residues M1-L49.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19059, 29749, 5535
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.