Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP4C Monoclonal Antibody (6B3)
GREENER_CHOICE

Catalog No. PIMA532946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532946 Supplier Invitrogen™ Supplier No. MA532946
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Purity is > 95% by SDS-PAGE.

The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP4C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP4C
Gene Accession No. P60510
Gene Alias 1110002D08Rik; AU016079; P PPX; pp4; PP4C; PPH3; Ppp4; PPP4C; PPX; PP-X; protein phosphatase 4 (formerly X), catalytic subunit; protein phosphatase 4 catalytic subunit; protein phosphatase 4, catalytic subunit; protein phosphatase X; protein phosphatase X, catalytic subunit; serine/threonine-protein phosphatase 4 catalytic subunit
Gene Symbols PPP4C
Host Species Mouse
Immunogen Recombinant full length Human PPP4C.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.