Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP4R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110211 Supplier Invitrogen™ Supplier No. PA5110211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NPSSSGQYFK EESKSSEEMS VENKNRTRDQ EAPEDVQVRP EDTPSDLSVS NSSVILENTM EDHAAEASGK PLGEISVPLD SSLLCTLSSE SHQEAASNEN DKKPGNYKSM LRPEVGTTSQ DSALLDQELY NSFHFWRTPL PEIDLDIELE QNSGGKPSPE GPEEESEGPV PSSPNITMAT RKELEEMIEN LEPHIDDPDV KAQVEVLSAA LRASSLDAHE ETISIEKRSD LQDELDINEL PNCKINQEDS VPLISDAVEN MDSTLHYIHS DSDLSNNSSF SPDEERRTKV QDVVPQALLD QYLSMTDPSR AQTVDTEIAK HCAY.

Regulatory subunit of serine/threonine-protein phosphatase 4. May play a role in regulation of cell division in renal glomeruli. The PPP4C-PPP4R1 PP4 complex may play a role in dephosphorylation and regulation of HDAC3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP4R1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP4R1
Gene Accession No. Q8K2V1, Q8TF05
Gene Alias 3110001J10Rik; MEG1; meg-1; PP4(Rmeg); PP4R1; Ppp4r1; protein phosphatase 4 regulatory subunit 1; protein phosphatase 4, regulatory subunit 1; serine/threonine phosphatase 4; serine/threonine-protein phosphatase 4 regulatory subunit 1
Gene Symbols PPP4R1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 324-647 of human PPP4R1 (NP_001035847.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70351, 9989
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.