Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP4R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674130
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-130 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-130 Supplier Invitrogen™ Supplier No. PA112397
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

PPP4R1 Polyclonal Antibody for Western Blot

Regulatory subunit of serine/threonine-protein phosphatase 4. May play a role in regulation of cell division in renal glomeruli. The PPP4C-PPP4R1 PP4 complex may play a role in dephosphorylation and regulation of HDAC3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP4R1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PPP4R1
Gene Accession No. Q8K2V1, Q8TF05, Q8VI02
Gene Alias 3110001J10Rik; MEG1; meg-1; PP4(Rmeg); PP4R1; Ppp4r1; protein phosphatase 4 regulatory subunit 1; protein phosphatase 4, regulatory subunit 1; serine/threonine phosphatase 4; serine/threonine-protein phosphatase 4 regulatory subunit 1
Gene Symbols PPP4R1
Host Species Sheep
Immunogen Synthetic peptide corresponding to residues 38-52 of the human protein phosphatase 4R1.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140943, 70351, 9989
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.