Learn More
PPP4R1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300837AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Regulatory subunit of serine/threonine-protein phosphatase 4. May play a role in regulation of cell division in renal glomeruli. The PPP4C-PPP4R1 PP4 complex may play a role in dephosphorylation and regulation of HDAC3.Specifications
| PPP4R1 | |
| Polyclonal | |
| Unconjugated | |
| PPP4R1 | |
| PP4(Rmeg), PP4R1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9989 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q8TF05 | |
| PPP4R1 | |
| Between 880 and C-term | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.