Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP4R1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106740 Supplier Invitrogen™ Supplier No. PA5106740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP4R1L.

May be a regulatory subunit of serine/threonine-protein phosphatase 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP4R1L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP4R1L
Gene Accession No. Q9P1A2
Gene Alias C20orf192; PPP4R1L; PPP4R1P1; PRO1085; protein phosphatase 4 regulatory subunit 1 like (pseudogene)
Gene Symbols PPP4R1L
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP4R1L(Accession Q9P1A2), corresponding to amino acid residues D387-W415.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55370
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.