Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP4R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531932 Supplier Invitrogen™ Supplier No. PA531932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, Jurkat, PPP4R2 transfected 293T lysate. Predicted reactivity: Mouse (88%), Rat (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a regulatory subunit of the serine/threonine-protein phosphatase 4 complex. In addition to being required for efficient DNA double strand break repair, this complex plays a role in organization of microtubules at centrosomes and processing of spliceosomal snRNPs. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP4R2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PPP4R2
Gene Accession No. Q9NY27
Gene Alias BE691708; C230060M08Rik; MGC131930; PP4R2; PPP4R2; protein phosphatase 4 regulatory subunit 2; protein phosphatase 4, regulatory subunit 2; SBBI57; Serine/threonine-protein phosphatase 4 regulatory subunit 2
Gene Symbols PPP4R2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP4R2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151987
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.