Learn More
PPP5C Mouse anti-Canine, Human, Rat, Clone: OTI5A8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500619

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.Specifications
PPP5C | |
Monoclonal | |
Unconjugated | |
PPP5C | |
PP5, PPP5, PPT | |
Mouse | |
Affinity Chromatography | |
RUO | |
5536, 612199, 65179 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5A8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P53041, P53042 | |
PPP5C | |
Full-length protein expressed in 293T cell transfected with human PPP5C expression vector | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.