Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP6C Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002883R
Description
PPP6C Polyclonal antibody specifically detects PPP6C in Human, Mouse samples. It is validated for Western BlotSpecifications
PPP6C | |
Polyclonal | |
Western Blot | |
EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
The immunogen recognized by this antibody maps to a region between residue 250 and the C-terminus (residue 305) of human Protein Phosphatase 6, Catalytic Subunit using the numbering given in entry NP_002712.1 (GeneID 5537). | |
0.1 mL | |
Cell Cycle and Replication, Signal Transduction | |
5537 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction