Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP6C Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | PPP6C |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PPP6C Polyclonal specifically detects PPP6C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PPP6C | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
5537 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: CYRCGNIASIMVFKDVNTREPKLFRAVPDSERVIPPRTTTPYFL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
PPP6C | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title