Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP6C Polyclonal Antibody

PPP6C Polyclonal Antibody
Supplier: Thermo Scientific PA528919
Description
PPP6C Polyclonal Antibody for Western Blot, IHC (P), ChIP
Specifications
| PPP6C | |
| Polyclonal | |
| Unconjugated | |
| Ppp6c | |
| 2310003C10Rik; ANKRD33; FLJ34236; PP6; PP6C; PPP6; PPP6C; Ppv; PP-V; protein phosphatase 6 catalytic subunit; protein phosphatase 6, catalytic subunit; protein phosphatase V; serine/threonine protein phosphatase catalytic subunit; serine/threonine-protein phosphatase 6 catalytic subunit; Serine/threonine-protein phosphatase 6 catalytic subunit, N-terminally processed; UNQ5837 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5537 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot | |
| 0.51 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O00743 | |
| Ppp6c | |
| Full length human PPP6C Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction