Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP6C Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen PPP6C
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-155-8596
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A300844AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

This gene encodes the catalytic subunit of protein phosphatase, a component of a signaling pathway regulating cell cycle progression. Splice variants encoding different protein isoforms exist. The pseudogene of this gene is located on chromosome X.
Specifications

Specifications

PPP6C
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
UNQ5837, ANKRD33, FLJ34236
Between 250 and C-term
Primary
4° C
PPP6C
Western Blot
Unconjugated
Rabbit
Human, Mouse
O00743, Q9CQR6
5537, 67857
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.