Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP6R1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560574
Description
Immunogen sequence: SALSANDAAL THELLALDVP NTMLDLFFHY VFNNFLHAQV EGCVSTMLSL GPPPDSSPET PIQNPVVKHL LQQCRLVERI LTSWEENDRV QCA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
Protein phosphatase regulatory subunits, such as SAPS1, modulate the activity of protein phosphatase catalytic subunits by restricting substrate specificity, recruiting substrates, and determining the intracellular localization of the holoenzyme. SAPS1 is a regulatory subunit for the protein phosphatase-6 catalytic subunit.
Specifications
PPP6R1 | |
Polyclonal | |
Unconjugated | |
Ppp6r1 | |
2010309P17Rik; AI836219; B430201G11Rik; D030074N20Rik; KIAA1115; LOW QUALITY PROTEIN: serine/threonine-protein phosphatase 6 regulatory subunit 1; MGC138185; MGC142003; mKIAA1115; Pp6r1; PPP6R1; protein phosphatase 6 regulatory subunit 1; protein phosphatase 6, regulatory subunit 1; RGD1311409; SAP190; SAPS domain family member 1; SAPS domain family, member 1; Saps1; Serine/threonine-protein phosphatase 6 regulatory subunit 1; Unknown (protein for MGC:128005) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22870 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPN7 | |
Ppp6r1 | |
Recombinant protein corresponding to Human PPP6R1. Recombinant protein control fragment (Product #RP-98953). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction