Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP6R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544275 Supplier Invitrogen™ Supplier No. PA544275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MFWKFDLHTS SHLDTLLERE DLSLPELLDE EDVLQECKVV NRKLLDFLLQ Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

Protein phosphatase regulatory subunits, such as SAPS1, modulate the activity of protein phosphatase catalytic subunits by restricting substrate specificity, recruiting substrates, and determining the intracellular localization of the holoenzyme. SAPS1 is a regulatory subunit for the protein phosphatase-6 catalytic subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP6R1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ppp6r1
Gene Accession No. Q9UPN7
Gene Alias 2010309P17Rik; AI836219; B430201G11Rik; D030074N20Rik; KIAA1115; LOW QUALITY PROTEIN: serine/threonine-protein phosphatase 6 regulatory subunit 1; MGC138185; MGC142003; mKIAA1115; Pp6r1; PPP6R1; protein phosphatase 6 regulatory subunit 1; protein phosphatase 6, regulatory subunit 1; RGD1311409; SAP190; SAPS domain family member 1; SAPS domain family, member 1; Saps1; Serine/threonine-protein phosphatase 6 regulatory subunit 1; Unknown (protein for MGC:128005)
Gene Symbols Ppp6r1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PPP6R1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22870
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.