Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP6R3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558405 Supplier Invitrogen™ Supplier No. PA558405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADQDDIGNVS FDRVSDINFT LNTNESGNIA LFEACCKERI QQFDDGGSDE EDIWEEKHIA FTPESQRRSS S Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

RMI1 is an essential component of the RMI complex, a complex that plays an important role in the processing of homologous recombination intermediates to limit DNA crossover formation in cells. RMI1 promotes TOP3A binding to double Holliday junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP6R3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP6R3
Gene Accession No. Q5H9R7
Gene Alias 4930528G08Rik; 9130026N02Rik; C11orf23; D19Bwg1430e; D19Ertd703e; DKFZp781E17107; DKFZp781E2374; DKFZp781O2362; FLJ11058; FLJ43065; KIAA1558; MGC125711; MGC125712; mKIAA1558; pluripotent cell-specific 3; Pp6r3; Ppcs3; PPP6R3; Ppp6r3-ps1; Pptcs3; protein phosphatase 6 regulatory subunit 3; protein phosphatase 6, regulatory subunit 3; RGD1307145; SAP190; Sapl; SAPLa; SAPS domain family member 3; SAPS domain family, member 3; SAPS domain family, member 3, pseudogene 1; SAPS3; Saps3-ps1; serine/threonine-protein phosphatase 6 regulatory subunit 3; sporulation-induced transcript 4-associated protein; sporulation-induced transcript 4-associated protein SAPL
Gene Symbols PPP6R3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP6R3. Recombinant protein control fragment (Product #RP-97300).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55291
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.