Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PPQ 102
Potent, voltage-independent CFTR chloride channel inhibitor
Specifications
Purity | >98% |
---|---|
Product Type | PPQ 102 |
Molecular Weight (g/mol) | 438.48 |
Inhibitors | PPQ 102 |
Description
Potent, voltage-independent CFTR chloride channel inhibitor (IC50 = 90 nM). Reduces the size and number of renal cysts in an embryonic kidney culture model of polycystic kidney disease. Also increases VEGF-A production from airway epithelial cells in vitro.Specifications
>98% | |
PPQ 102 | |
438.48 | |
PPQ 102 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title