Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558420 Supplier Invitrogen™ Supplier No. PA558420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASPHPKHKVS ALVQSPQMKA LACVSAEGVT VEEPASERLK PETQETRPRE KPPLPATKAV PTPRQSTVPK LPAVHPARLR KLSFLPTPRT QGSEDVVQ Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 76%.

The protein encoded by this gene is similar to PPAR-gamma coactivator 1 (PPARGC1/PGC-1), a protein that can activate mitochondrial biogenesis in part through a direct interaction with nuclear respiratory factor 1 (NRF1). This protein has been shown to interact with NRF1. It is thought to be a functional relative of PPARGC1 that activates mitochondrial biogenesis through NRF1 in response to proliferative signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPRC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPRC1
Gene Accession No. Q5VV67
Gene Alias A930018M19; BC013720; KIAA0595; mKIAA0595; peroxisome proliferative activated receptor, gamma, coactivator-related 1; peroxisome proliferator-activated receptor gamma coactivator-related protein 1; peroxisome proliferator-activated receptor gamma, coactivator-related 1; PGC-1-related coactivator; PPRC1; PRC; RP11-302K17.6
Gene Symbols PPRC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPRC1. Recombinant protein control fragment (Product #RP-97117).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23082
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.